Order Entry
Czech Republic
ContactUsLinkComponent
(S)-2,2'-Dimethoxy-1,1'-binaphthalene-3,3'-diboronic acid 98%
(S)-2,2'-Dimethoxy-1,1'-binaphthalene-3,3'-diboronic acid 98%
Catalog # APOSOR360578-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-2,2'-Dimethoxy-1,1'-binaphthalene-3,3'-diboronic acid 98%
Catalog # APOSOR360578-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :402.02 g/mol
Storage Temperature:Ambient
CAS Number:220204-00-2
Formula:C₂₂H₂₀B₂O₆
MDL Number:MFCD09027249