Order Entry
Czech Republic
ContactUsLinkComponent
 
4-(Morpholinomethyl)phenylboronic acid 98%
Catalog # APOSOR360236-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Morpholinomethyl)phenylboronic acid 98%
Catalog # APOSOR360236-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.06 g/mol
Melting Pt:81…85 °C
Storage Temperature:Refrigerator
CAS Number:279262-23-6
Formula:C₁₁H₁₆BNO₃
MDL Number:MFCD06801707