Order Entry
Czech Republic
ContactUsLinkComponent
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine 95+%
Irritant
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine 95+%
Catalog # APOSOR360160-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine 95+%
Catalog # APOSOR360160-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :245.0853 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1093819-50-1
Formula:C₁₂H₁₆BN₃O₂
MDL Number:MFCD09832892