Order Entry
Czech Republic
ContactUsLinkComponent
3-Boc-9-oxo-3-azaspiro[5.5]undecane 97%
3-Boc-9-oxo-3-azaspiro[5.5]undecane 97%
Catalog # APOSOR350440-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Boc-9-oxo-3-azaspiro[5.5]undecane 97%
Catalog # APOSOR350440-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :267.37 g/mol
Storage Temperature:Ambient
CAS Number:873924-08-4
Formula:C₁₅H₂₅NO₃
MDL Number:MFCD12198558