(S)-4-(4-Aminobenzyl)oxazolidin-2-one
Catalog # APOSOR350358-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:192.22 g/mol
Storage Temperature:Ambient
CAS Number:152305-23-2
:340041-89-6
Formula:C₁₀H₁₂N₂O₂
MDL Number:MFCD03411476