(S)-2-(2-Hydroxyethyl)piperidine hydrochloride
Catalog # APOSOR350353-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:165.6610 g/mol
Storage Temperature:Ambient
CAS Number:786684-21-7
Formula:C₇H₁₆ClNO
MDL Number:MFCD09265049