Pyridoxal-5'-phosphate monohydrate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:41468-25-1
:
Formula:C₈H₁₀NO₆P·1H₂O
MDL Number:MFCD00149414
:265.17 g/mol (247.15 anhy)
Storage Temperature:Freezer
Frequently Bought Together