Order Entry
Czech Republic
ContactUsLinkComponent
2-Methyl-3-tetrahydrothiophenone
2-Methyl-3-tetrahydrothiophenone
Catalog # APOSOR350261-25G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-3-tetrahydrothiophenone
Catalog # APOSOR350261-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :116.18 g/mol
Storage Temperature:Ambient
CAS Number:13679-85-1
Formula:C₅H₈OS
MDL Number:MFCD00078280