Order Entry
Czech Republic
ContactUsLinkComponent
1-Methyl-2-pyrrolecarboxaldehyde
1-Methyl-2-pyrrolecarboxaldehyde
Catalog # APOSOR350219-100G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-2-pyrrolecarboxaldehyde
Catalog # APOSOR350219-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :109.13 g/mol
Storage Temperature:Ambient
CAS Number:1192-58-1
Formula:C₆H₇NO
MDL Number:MFCD00003087