Order Entry
Czech Republic
ContactUsLinkComponent
Methyl cyclohexanecarboxylate > 98%
Flammable
Irritant
Methyl cyclohexanecarboxylate > 98%
Catalog # APOSOR350217-25G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
Methyl cyclohexanecarboxylate > 98%
Catalog # APOSOR350217-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :142.2 g/mol
Boiling Pt:183 °C
Density:0.98 g/cm³ (25 °C)
Flash Pt:60 °C
UN:3272
ADR:3,III
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:4630-82-4
Formula:C₈H₁₄O₂
MDL Number:MFCD00001458