Order Entry
Czech Republic
ContactUsLinkComponent
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate
Catalog # APOSOR350152-25G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate
Catalog # APOSOR350152-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :254.2800 g/mol
Storage Temperature:Ambient
CAS Number:144689-94-1
Formula:C₁₂H₁₈N₂O₄
MDL Number:MFCD08062365