
4,4'-Dimethoxytrityl chloride 98%
:40615-36-9
:
:C₂₁H₁₉ClO₂
:MFCD00008409
:338.8 g/mol
:119…123 °C
:GHS07
:Ambient
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
: VWR Chemicals
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
(±)-2-Butanol ≥99% analytical reagent
: VWR Chemicals