Order Entry
Czech Republic
ContactUsLinkComponent
 
7-Methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one 95+%
Catalog # APOSOR346572-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one 95+%
Catalog # APOSOR346572-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.2 g/mol
Storage Temperature:Ambient
CAS Number:700-52-7
Formula:C₇H₆N₂OS
MDL Number:MFCD00552712