Order Entry
Czech Republic
ContactUsLinkComponent
1-[6-(3-Methylphenyl)pyridin-3-yl]ethanone 95+%
1-[6-(3-Methylphenyl)pyridin-3-yl]ethanone 95+%
Catalog # APOSOR346548-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[6-(3-Methylphenyl)pyridin-3-yl]ethanone 95+%
Catalog # APOSOR346548-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211.27 g/mol
Storage Temperature:Ambient
CAS Number:1217035-58-9
Formula:C₁₄H₁₃NO
MDL Number:MFCD12404919