Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone 95+%
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone 95+%
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone 95+%
APOSOR346547-1G
:
Apollo Scientific
:
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone 95+%
APOSOR346547-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
227.27 g/mol
:
Ambient
:
1215576-19-4
:
C₁₄H₁₃NO₂
:
MFCD12404914