
4-Methyl-1H-indole-2-carboxylic acid methyl ester
Catalog # APOSOR346108-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:189.22 g/mol
Melting Pt:117…120 °C
:GHS07
Storage Temperature:Ambient
CAS Number:136831-13-5
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD04966966