Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(1-Methyl-1H-pyrazol-3-yl)phenylamine
2-(1-Methyl-1H-pyrazol-3-yl)phenylamine
2-(1-Methyl-1H-pyrazol-3-yl)phenylamine
APOSOR346099-500MG
:
Apollo Scientific
:
2-(1-Methyl-1H-pyrazol-3-yl)phenylamine
APOSOR346099-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
173.22 g/mol
:
59…60 °C
:
GHS07
:
Ambient
:
87488-80-0
:
C₁₀H₁₁N₃
:
MFCD11052424