

2-Chloro-4-iodo-3-methylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:153034-88-9
:
Formula:C₆H₅ClIN
MDL Number:MFCD03095293
:253.47 g/mol
Melting Pt:101…106 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together