Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
C-(2,3-Dihydrobenzo[1,4]dioxin-2-yl)methylamine
C-(2,3-Dihydrobenzo[1,4]dioxin-2-yl)methylamine
C-(2,3-Dihydrobenzo[1,4]dioxin-2-yl)methylamine
APOSOR3418-5G
:
Apollo Scientific
:
C-(2,3-Dihydrobenzo[1,4]dioxin-2-yl)methylamine
APOSOR3418-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
165.19 g/mol
:
105…110 °C/3mmHg
:
1.1679 g/cm³ (25 °C)
:
224-671-6
:
GHS05
:
GHS07
:
Ambient
:
4442-59-5
:
C₉H₁₁NO₂
:
MFCD00203985