

2-Chloro-4-nitrophenyl isocyanate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40397-95-3
:
Formula:C₇H₃ClN₂O₃
MDL Number:MFCD00037063
:198.57 g/mol
UN:2811
ADR:6.1,III
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together