
Methyl-5-bromothiazole-4-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:913836-22-3
:
Formula:C₅H₄BrNO₂S
MDL Number:MFCD08275713
:222.07 g/mol
Boiling Pt:276 °C
Melting Pt:98…101 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together