Order Entry
Czech Republic
ContactUsLinkComponent
(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol 95%
(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol 95%
Catalog # APOSOR33634-1G
Supplier:  Apollo Scientific
CAS Number:  
(4,5-Dibromo-1-methyl-1H-imidazol-2-yl)methanol 95%
Catalog # APOSOR33634-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :269.93 g/mol
Storage Temperature:Ambient
CAS Number:881997-90-6
Formula:C₅H₆Br₂N₂O
MDL Number:MFCD12025921