Order Entry
Czech Republic
ContactUsLinkComponent
2-{[(4-Chlorophenyl)methyl]amino}-2-methylpropanenitrile 97%
2-{[(4-Chlorophenyl)methyl]amino}-2-methylpropanenitrile 97%
Catalog # APOSOR33597-1G
Supplier:  Apollo Scientific
CAS Number:  
2-{[(4-Chlorophenyl)methyl]amino}-2-methylpropanenitrile 97%
Catalog # APOSOR33597-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :208.69 g/mol
Storage Temperature:Ambient
CAS Number:1018265-80-9
Formula:C₁₁H₁₃ClN₂
MDL Number:MFCD10014479