Order Entry
Czech Republic
ContactUsLinkComponent
5-(3-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione 95%
5-(3-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione 95%
Catalog # APOSOR33581-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(3-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione 95%
Catalog # APOSOR33581-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.16 g/mol
Storage Temperature:Ambient
CAS Number:333771-24-7
Formula:C₉H₈BrN₃S
MDL Number:MFCD01816005