Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl-3-methoxy-4-(pyrrolidin-1-ylmethyl)benzoate 97%
  APOSOR33562-1G
 :  
Methyl-3-methoxy-4-(pyrrolidin-1-ylmethyl)benzoate 97%
  APOSOR33562-1G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

 :
 :249.31 g/mol
 :Ambient
 :193964-75-9
 :C₁₄H₁₉NO₃
 :MFCD22126102