Order Entry
Czech Republic
ContactUsLinkComponent
6-(Dimethylamino)-2-methyl-4-pyrimidinol 95%
6-(Dimethylamino)-2-methyl-4-pyrimidinol 95%
Catalog # APOSOR33489-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(Dimethylamino)-2-methyl-4-pyrimidinol 95%
Catalog # APOSOR33489-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :153.19 g/mol
Storage Temperature:Ambient
CAS Number:83724-17-8
Formula:C₇H₁₁N₃O
MDL Number:MFCD11840974