Order Entry
Czech Republic
ContactUsLinkComponent
[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol 95%
[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol 95%
Catalog # APOSOR33488-1G
Supplier:  Apollo Scientific
CAS Number:  
[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol 95%
Catalog # APOSOR33488-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.3 g/mol
Storage Temperature:Ambient
CAS Number:889954-12-5
Formula:C₁₆H₁₆O₃
MDL Number:MFCD06802587