Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone
1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone 95%
1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone 95%
APOSOR33474-1G
:
Apollo Scientific
:
1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone 95%
APOSOR33474-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
239.08 g/mol
:
Ambient
:
99661-02-6
:
C₁₀H₇BrO₂
:
MFCD09972223