Order Entry
Czech Republic
ContactUsLinkComponent
2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime 95%
2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime 95%
Catalog # APOSOR33462-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime 95%
Catalog # APOSOR33462-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.2 g/mol
Storage Temperature:Ambient
CAS Number:1017782-44-3
Formula:C₁₀H₉N₃O
MDL Number:MFCD09152718