
2-(2-Propynyloxy)benzenecarbaldehyde 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29978-83-4
:
Formula:C₁₀H₈O₂
MDL Number:MFCD02380378
:160.17 g/mol
Melting Pt:70 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together