Methyl-2-hydroxy-4-(1H-pyrrol-1-yl)benzenecarboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:454473-64-4
:
Formula:C₁₂H₁₁NO₃
MDL Number:MFCD02083001
:217.23 g/mol
Storage Temperature:Ambient
Frequently Bought Together