Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid 95%
2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid 95%
Catalog # APOSOR33205-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid 95%
Catalog # APOSOR33205-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.66 g/mol
Storage Temperature:Ambient
CAS Number:220461-68-7
Formula:C₁₁H₉ClN₂O₂
MDL Number:MFCD08056672