Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate 95%
Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate 95%
Catalog # APOSOR33202-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate 95%
Catalog # APOSOR33202-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.04 g/mol
Storage Temperature:Ambient
CAS Number:1232838-31-1
Formula:C₆H₇BrN₂O₂
MDL Number:MFCD08056667