Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(3-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol
3-(3-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol 95%
3-(3-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol 95%
APOSOR33116-1G
:
Apollo Scientific
:
3-(3-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol 95%
APOSOR33116-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
221.65 g/mol
:
Ambient
:
886360-72-1
:
C₁₀H₈ClN₃O
:
MFCD04117818