Order Entry
Czech Republic
ContactUsLinkComponent
2-(Methylsulphanyl)-5-phenylpyrimidine 95%
2-(Methylsulphanyl)-5-phenylpyrimidine 95%
Catalog # APOSOR33092-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Methylsulphanyl)-5-phenylpyrimidine 95%
Catalog # APOSOR33092-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :202.28 g/mol
Storage Temperature:Ambient
CAS Number:31408-20-5
Formula:C₁₁H₁₀N₂S
MDL Number:MFCD00204407