Order Entry
Czech Republic
ContactUsLinkComponent
1-[2,3-Dichloro-4-(ethylsulphonyl)phenyl]-1-ethanone 95%
1-[2,3-Dichloro-4-(ethylsulphonyl)phenyl]-1-ethanone 95%
Catalog # APOSOR33091-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2,3-Dichloro-4-(ethylsulphonyl)phenyl]-1-ethanone 95%
Catalog # APOSOR33091-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :281.16 g/mol
Storage Temperature:Ambient
CAS Number:338982-46-0
Formula:C₁₀H₁₀Cl₂O₃S
MDL Number:MFCD01935964