4-(4-Chlorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:39561-83-6
:
Formula:C₁₁H₆ClNO₂
MDL Number:MFCD00099688
:219.63 g/mol
Storage Temperature:Ambient
Frequently Bought Together