Order Entry
Czech Republic
ContactUsLinkComponent
2-(1H-Pyrazol-3-yl)pyrazine 95%
2-(1H-Pyrazol-3-yl)pyrazine 95%
Catalog # APOSOR32945-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-(1H-Pyrazol-3-yl)pyrazine 95%
Catalog # APOSOR32945-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :146.15 g/mol
Storage Temperature:Ambient
CAS Number:111781-54-5
Formula:C₇H₆N₄
MDL Number:MFCD03618495