
[3-(4-Methoxyphenyl)-1H-pyrazol-1-yl]acetic acid
Catalog # APOSOR3289-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:232.24 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:959584-25-9
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD00665860