Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-4-Aminocyclohexanecarboxylic acid
cis-4-Aminocyclohexanecarboxylic acid
cis-4-Aminocyclohexanecarboxylic acid
APOSOR3286-1G
:
Apollo Scientific
:
cis-4-Aminocyclohexanecarboxylic acid
APOSOR3286-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
143.19 g/mol
:
299…301 °C
:
GHS07
:
Ambient
:
3685-23-2
:
C₇H₁₃NO₂
:
MFCD00191730