Order Entry
Czech Republic
ContactUsLinkComponent
4-(4-Chlorophenyl)-3-methyl-1H-pyrazole 95%
4-(4-Chlorophenyl)-3-methyl-1H-pyrazole 95%
Catalog # APOSOR32692-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(4-Chlorophenyl)-3-methyl-1H-pyrazole 95%
Catalog # APOSOR32692-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.65 g/mol
Storage Temperature:Ambient
CAS Number:667400-41-1
Formula:C₁₀H₉ClN₂
MDL Number:MFCD00179386