0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole
2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole 95%
2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole 95%
APOSOR32627-1G
:
Apollo Scientific
:
2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole 95%
APOSOR32627-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
229.67 g/mol
:
Ambient
:
143426-40-8
:
C₁₂H₈ClN₃
:
MFCD12025828