Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile 95%
2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile 95%
Catalog # APOSOR32606-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile 95%
Catalog # APOSOR32606-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (2Z)-2-(4-Chlorophenyl)-3-(dimethylamino)prop-2-enenitrile
 :
 :206.68 g/mol
Storage Temperature:Ambient
CAS Number:40373-88-4
Formula:C₁₁H₁₁ClN₂
MDL Number:MFCD00138415