Order Entry
Czech Republic
ContactUsLinkComponent
1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone 95%
1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone 95%
Catalog # APOSOR32427-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Bromophenyl)-3,3-dimethyl-2-azetanone 95%
Catalog # APOSOR32427-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.13 g/mol
Storage Temperature:Ambient
CAS Number:338392-04-4
Formula:C₁₁H₁₂BrNO
MDL Number:MFCD00793408