
3-(Trimethoxysilyl)propyl methacrylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2530-85-0
:
Formula:C₁₀H₂₀O₅Si
MDL Number:MFCD00008593
:248.35 g/mol
Boiling Pt:190 °C
Density:1.045 g/cm³ (25 °C)
Flash Pt:92 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together