Order Entry
Czech Republic
ContactUsLinkComponent
5-Formyl-2-methoxyphenyl Acetate 97%
5-Formyl-2-methoxyphenyl Acetate 97%
Catalog # APOSOR322864-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Formyl-2-methoxyphenyl Acetate 97%
Catalog # APOSOR322864-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :194.19 g/mol
Storage Temperature:Ambient
CAS Number:881-57-2
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00266441