Order Entry
Czech Republic
ContactUsLinkComponent
2-(1-Methylhydrazino)-4,5-dihydro-1H-imidazole hydrobromide
2-(1-Methylhydrazino)-4,5-dihydro-1H-imidazole hydrobromide
Catalog # APOSOR322478-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(1-Methylhydrazino)-4,5-dihydro-1H-imidazole hydrobromide
Catalog # APOSOR322478-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.07 g/mol
Storage Temperature:Ambient
CAS Number:55959-80-3
Formula:C₄H₁₁BrN₄
MDL Number:MFCD00067859