Methyl-4-bromo-2-methoxybenzoate
Catalog # APOSOR322451-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:245.07 g/mol
Storage Temperature:Ambient
CAS Number:139102-34-4
Formula:C₉H₉BrO₃
MDL Number:MFCD06797867