5-Bromo-2-hydroxy-3-methoxybenzyl alcohol, Technical Grade
Catalog # APOSOR32240-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.06
Storage Temperature:Ambient
CAS Number:35090-64-3
Formula:C₈H₉BrO₃
MDL Number:MFCD02081986