Order Entry
Czech Republic
ContactUsLinkComponent
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
Catalog # APOSOR322293-250MG
Supplier:  Apollo Scientific
CAS Number:  
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
Catalog # APOSOR322293-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :330.39 g/mol
Storage Temperature:Ambient
CAS Number:175166-49-1
Formula:C₂₁H₁₈N₂O₂
MDL Number:MFCD00799561